Potassium chlorate ≥99.0% ACS

Supplier: BeanTown Chemical

225740-2KG 225740-500G
BT225740-2KGEA 187 USD
BT225740-2KG BT225740-500G
Potassium chlorate ≥99.0% ACS
Potassium chlorate

CAS: 3811-04-9; EC No: 223-289-7; MDL No: MFCD00011361; RTECS: FO0350000 UN No: UN1485; Haz Class: 5.1; Packing Group: II Crystalline; Linear Formula: KClO3; MW: 122.55 Melting Point: 356° Density (g/mL): 2.32 Hygroscopic


  • RTECS: FO0350000
  • Hygroscopic
  • Crystalline

Formula: KClO₃
MW: 122.55 g/mol
Melting Pt: 356 °C
Density: 2.32 g/mL
MDL Number: MFCD00011361
CAS Number: 3811-04-9
EINECS: 223-289-7
UN: 1485
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR