Lead(II) acetate trihydrate 99.0-103.0% ACS

Supplier: BeanTown Chemical

Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate

225295-100G 225295-2KG 225295-500G
BT225295-100GEA 41.94 USD
BT225295-100G BT225295-2KG BT225295-500G
Lead(II) acetate trihydrate 99.0-103.0% ACS
Lead (II) acetate trihydrate

CAS: 6080-56-4; EC No: 206-104-4; MDL No: MFCD00150023; RTECS: OF8050000 UN No: UN1616; Haz Class: 6.1; Packing Group: III Granular; Linear Formula: Pb(OOCCH3)2·3H2O; MW: 379.33 Melting Point: 75° (decomposes)


  • RTECS: OF8050000
  • Granular

Formula: (H₃CCOO)₂Pb·3H₂O
MW: 379.34 g/mol
Melting Pt: 75 °C (dec.)
MDL Number: MFCD00150023
CAS Number: 6080-56-4
EINECS: 206-104-4
UN: 1616
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351511a44fbb598a2d7fa1fa4ab395427b6168f1&vid=67e858e5f0dc4d04dd796541b3c0ad5aa73f8dc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0-103.0%
Insoluble matter ≤0.01%
Chloride (Cl) ≤5ppm
Copper (Cu) ≤0.002%
Nitrate/Nitrite ≤0.005%
Iron (Fe) ≤0.001%
Sodium (Na) ≤0.005%
Potassium (K) ≤0.005%
Calcium (Ca) ≤0.005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR