Tetramethoxysilane ≥98%
Supplier: BeanTown Chemical
Synonyms:
Tetramethyl orthosilicate
224600-100G
224600-25G
224600-500G
BT224600-100GEA
89.49
USD
BT224600-100G
BT224600-25G
BT224600-500G
Tetramethoxysilane ≥98%
Tetramethoxysilane
CAS: 681-84-5; EC No: 211-656-4; MDL No: MFCD00008341; RTECS: VV9800000
UN No: UN2606; Haz Class: 6.1 (3); Packing Group: I
Liquid; Linear Formula: Si(OCH3)4; Molecular Formula: C4H12O4Si; MW: 152.22
Melting Point: -4°; Boiling Point: 121-122°; Flash point: 26°C (79°F)
Density (g/mL): 1.023; Refractive Index: 1.368
Moisture Sensitive
- RTECS: VV9800000
- Refractive index: 1.368
- Moisture Sensitive
- Liquid
Formula:
C₄H₁₂O₄Si MW: 152.22 g/mol Boiling Pt: 121 Melting Pt: -2 Density: 1.023 g/mL Flash Pt: 26 °C (79 °F) |
MDL Number:
MFCD00008341 CAS Number: 681-84-5 EINECS: 211-656-4 UN: 2606 ADR: 6.1 I |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...