Trifluoromethanesulfonic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
Trifluoromethanesulphonic acid, Trifluoromethanesulfonic acid
224535-10G
224535-250G
224535-50G
BT224535-10GEA
29.78
USD
BT224535-10G
BT224535-250G
BT224535-50G
Trifluoromethanesulfonic acid ≥98%
Trifluoromethanesulphonic acid
CAS: 1493-13-6; EC No: 216-087-5; MDL No: MFCD00007514; RTECS: PB2771000
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: CF3SO3H; MW: 150.07
Melting Point: -40°; Boiling Point: 162°
Density (g/mL): 1.696; Refractive Index: 1.327
Hygroscopic
- RTECS: PB2771000
- Refractive index: 1.327
- Hygroscopic
- Liquid
Formula:
CHF₃O₃S MW: 150.08 g/mol Boiling Pt: 162 °C Melting Pt: –40 °C Density: 1.696 g/mL |
MDL Number:
MFCD00007514 CAS Number: 1493-13-6 EINECS: 216-087-5 UN: 3265 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...