Nickel ≥99.5%, soluble -325 mesh, Thickness 0.37 µm
Supplier: BeanTown Chemical
224150-1KG
224150-250G
BT224150-1KGEA
141
USD
BT224150-1KG
BT224150-250G
Nickel ≥99.5%, soluble -325 mesh, Thickness 0.37 µm
Nickel
CAS: 7440-02-0; EC No: 231-111-4; MDL No: MFCD00011137; RTECS: QR5950000
UN No: UN3089; Haz Class: 4.1; Packing Group: II
Solid; Molecular Formula: Ni; MW: 58.69
Melting Point: 1453°; Boiling Point: 2732°
Density (g/mL): 8.9
- RTECS: QR5950000
- Solid
Formula:
Ni Boiling Pt: 2732 °C Melting Pt: 1453 °C Density: 8.9 g/mL |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1,II |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...