6-Bromocaproic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 6-Bromohexanoic acid

224115-100G 224115-25G 224115-500G
BT224115-100GEA 182 USD
BT224115-100G BT224115-25G BT224115-500G
6-Bromocaproic acid ≥98%
6-Bromocaproic acid

CAS: 4224-70-8; EC No: 224-176-5; MDL No: MFCD00004422 UN No: UN3261; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: Br(CH2)5CO2H; Molecular Formula: C6H11BrO2; MW: 195.05 Melting Point: 32-34°; Boiling Point: 165-170°/20 mmHg; Flash point: 95°C (203°F)


  • Crystalline

Formula: BrH₂C(CH₂)₄COOH
MW: 195.06 g/mol
Boiling Pt: 165
Melting Pt: 32…34 °C
Flash Pt: 95 °C (203 °F)
MDL Number: MFCD00004422
CAS Number: 4224-70-8
EINECS: 224-176-5
UN: 3261
ADR: 8 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR