Iridium, black ≥99.95%
Supplier: BeanTown Chemical
223290-10G
223290-2G
223290-500MG
BT223290-10GEA
4330
USD
BT223290-10G
BT223290-2G
BT223290-500MG
Iridium, black ≥99.95%
Iridium
CAS: 7439-88-5; EC No: 231-095-9; MDL No: MFCD00011062
UN No: UN3089; Haz Class: 4.1; Packing Group: III
Solid, S.A. 30-40 m2/g ; Molecular Formula: Ir; MW: 192.22
Melting Point: 2450°; Boiling Point: 4130°
Density (g/mL): 22.65
- Solid, S.A. 30-40 m²/g
Formula:
Ir Boiling Pt: 4130 °C Melting Pt: 2450 °C Density: 22.65 g/mL |
MDL Number:
MFCD00011062 CAS Number: 7439-88-5 EINECS: 231-095-9 UN: 3089 ADR: 4.1,III |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...