Manganese ≥99% (trace metals basis), powder -325 mesh

Supplier: BeanTown Chemical

223250-1KG 223250-250G
BT223250-1KGEA 128.38 USD
BT223250-1KG BT223250-250G
Manganese ≥99% (trace metals basis), powder -325 mesh
Manganese

CAS: 7439-96-5; EC No: 231-105-1; MDL No: MFCD00011111 UN No: UN3089; Haz Class: 4.1; Packing Group: III Powder, apparent density 2.6-3.5g/cm3 ; Molecular Formula: Mn; MW: 54.94 Melting Point: 1244°; Boiling Point: 1962° Density (g/mL): 7.3


  • Powder, apparent density 2.6-3.5g/cm3

Formula: Mn
Boiling Pt: 1962 °C
Melting Pt: 1244 °C
Density: 7.3 g/mL
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1
UN: 3089
ADR: 4.3,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR