(S)-(+)-4-Penten-2-ol ≥97%
Supplier: BeanTown Chemical
Synonyms:
(S)-(+)-1-Penten-4-ol, (S)-(+)-Allyl methyl carbinol, (S)-Pent-4-en-2-ol
223175-1G
223175-5G
BT223175-1GEA
153
USD
BT223175-1G
BT223175-5G
(S)-(+)-4-Penten-2-ol ≥97%
(S)-(+)-4-Penten-2-ol
CAS: 55563-79-6; MDL No: MFCD03427200
UN No: UN1987; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C5H10O; MW: 86.13
Boiling Point: 115-116°; Flash point: 30°C (86°F)
Density (g/mL): 0.837; Refractive Index: 1.424; Optical Rotation: [α]20/D +5.0°, c = 1% in chloroform
- Optical rotation: [α]20/D +5.0°, c = 1% in chloroform
- Refractive index: 1.424
- Liquid
Formula:
C₅H₁₀O Boiling Pt: 115…116 °C Density: 0.837 g/mL Flash Pt: 30 °C (86 °F) |
MDL Number:
MFCD03427200 CAS Number: 55563-79-6 UN: 1987 ADR: 3,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...