Chromium(II) chloride, anhydrous ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Chromium dichloride

223155-1G 223155-25G 223155-5G
BT223155-1GEA 43 USD
BT223155-1G BT223155-25G BT223155-5G
Chromium(II) chloride, anhydrous ≥99.9% (trace metals basis)
Chromium (II) chloride

CAS: 10049-05-5; EC No: 233-163-3; MDL No: MFCD00010947; RTECS: GB5250000 UN No: UN3260; Haz Class: 8; Packing Group: III -80 Mesh Powder; Linear Formula: CrCl2; MW: 122.90 Melting Point: 824° Density (g/mL): 2.9 Air Sensitive, Moisture Sensitive


  • RTECS: GB5250000
  • Air Sensitive, Moisture Sensitive
  • -80 Mesh Powder

Formula: Cl₂Cr
MW: 122.9 g/mol
Melting Pt: 824 °C
Density: 2.9 g/mL
MDL Number: MFCD00010947
CAS Number: 10049-05-5
EINECS: 233-163-3
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR