Zinc diethyldithiocarbamate

Supplier: BeanTown Chemical

Synonyms: Zinc dethyldithiocarbamate, Diethyldithiocarbamic Acid Zinc Salt

222380-2KG 222380-500G
BT222380-2KGEA 74 USD
BT222380-2KG BT222380-500G
Zinc diethyldithiocarbamate
Zinc diethyldithiocarbamate

CAS: 14324-55-1; EC No: 238-270-9; MDL No: MFCD00064798; RTECS: ZH0350000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: [(CH3CH2)2NCS2]2Zn; Molecular Formula: C10H20N2S4Zn ; MW: 361.93 Melting Point: 178-181°; Boiling Point: 301°; Flash point: 204°C (400°F) Density (g/mL): 1.48


  • RTECS: ZH0350000
  • Powder

Formula: C₅H₁₁NS₂
MW: 361.90 g/mol
Boiling Pt: 301 °C
Melting Pt: 178…181 °C
Density: 1.48 g/mL
Flash Pt: 204 °C (400 °F)
MDL Number: MFCD00064798
CAS Number: 14324-55-1
EINECS: 238-270-9
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR