2,4,6-Tri-tert-butylaniline ≥95%
Supplier: BeanTown Chemical
Synonyms:
2,4,6-Tris(tert-butyl)aniline
221685-1G
221685-5G
BT221685-1GEA
50
USD
BT221685-1G
BT221685-5G
2,4,6-Tri-tert-butylaniline ≥95%
2,4,6-Tri-tert-butylaniline
CAS: 961-38-6; EC No: 213-507-9; MDL No: MFCD00011645
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C18H31N; MW: 261.45
Melting Point: 145-147°
Light Sensitive
- Light Sensitive
- Crystalline
Formula:
C₁₈H₃₁N Melting Pt: 145…147 °C |
MDL Number:
MFCD00011645 CAS Number: 961-38-6 EINECS: 213-507-9 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...