Diphenyl chlorophosphate ≥97%

Supplier: BeanTown Chemical

Synonyms: Diphenyl phosphorochloridate

221420-1KG 221420-250G 221420-50G
BT221420-1KGEA 251 USD
BT221420-1KG BT221420-250G BT221420-50G
Diphenyl chlorophosphate ≥97%
Diphenyl chlorophosphate

CAS: 2524-64-3; EC No: 219-759-6; MDL No: MFCD00003030 UN No: UN3265; Haz Class: 8; Packing Group: III Liquid; Linear Formula: (C6H5O)2POCl; Molecular Formula: C12H10ClO3P; MW: 268.64 Boiling Point: 314-316°/272 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.296; Refractive Index: 1.55 Moisture Sensitive


  • Refractive index: 1.55
  • Moisture Sensitive
  • Liquid

Formula: C₁₂H₁₀ClO₃P
MW: 268.64 g/mol
Boiling Pt: 314…316 °C/272 mmHg
Density: 1.296 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00003030
CAS Number: 2524-64-3
EINECS: 219-759-6
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR