Lanthanum ≥99.9% (trace rare earth metals basis), pieces

Supplier: BeanTown Chemical

221030-100G 221030-25G
BT221030-100GEA 265 USD
BT221030-100G BT221030-25G
Lanthanum ≥99.9% (trace rare earth metals basis), pieces
Lanthanum

CAS: 7439-91-0; EC No: 231-099-0; MDL No: MFCD00011066 UN No: UN3208; Haz Class: 4.3; Packing Group: II Solid, packaged under mineral oil; Molecular Formula: La; MW: 138.91 Melting Point: 920°; Boiling Point: 3464° Density (g/mL): 6.19


  • Solid, packaged under mineral oil

Formula: La
Boiling Pt: 3464 °C
Melting Pt: 920 °C
Density: 6.19 g/mL
MDL Number: MFCD00011066
CAS Number: 7439-91-0
EINECS: 231-099-0
UN: 3208
ADR: 4.3,II

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR