2,5-Dimercapto-1,3,4-thiadiazole ≥98%
Supplier: BeanTown Chemical
Synonyms:
1,3,4-Thiadiazole-2,5-dithiol
220145-250G
220145-50G
BT220145-250GEA
178
USD
BT220145-250G
BT220145-50G
2,5-Dimercapto-1,3,4-thiadiazole ≥98%
2,5-Dimercapto-1,3,4-thiadiazole
CAS: 1072-71-5; EC No: 214-014-1; MDL No: MFCD00003103; RTECS: XI3850000
UN No: UN3335; Haz Class: 9; Packing Group: III
Powder; Molecular Formula: C2H2N2S3; MW: 150.25
Melting Point: 162° (decomposes)
- RTECS: XI3850000
- Powder
Formula:
C₂H₂N₂S₃ MW: 150.25 g/mol Melting Pt: 162 °C (dec.) |
MDL Number:
MFCD00003103 CAS Number: 1072-71-5 EINECS: 214-014-1 ADR: 9 III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...