Sodium borohydride ≥98%, granular

Supplier: BeanTown Chemical

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

219965-100G 219965-500G
BT219965-100GEA 102 USD
BT219965-100G BT219965-500G
Sodium borohydride ≥98%, granular
Sodium borohydride

CAS: 16940-66-2; EC No: 241-004-4; MDL No: MFCD00003518; RTECS: ED3325000 UN No: UN1426; Haz Class: 4.3; Packing Group: I Mesh Granular; Linear Formula: NaBH4; MW: 37.83 Melting Point: 300° (decomposes) Density (g/mL): 1.074 Air Sensitive, Moisture Sensitive


  • RTECS: ED3325000
  • Air Sensitive, Moisture Sensitive
  • Mesh Granular

Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400 (dec.)
Density: 1.074 g/mL
Flash Pt: 69
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3 I
Merck Index: 13,08664

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR