Chloro(dimethyl)(phenyl)silane ≥97%
Supplier: BeanTown Chemical
Synonyms:
Chlorodimethylphenylsilane
218885-100G
218885-25G
218885-5G
BT218885-100GEA
356
USD
BT218885-100G
BT218885-25G
BT218885-5G
Chloro(dimethyl)(phenyl)silane ≥97%
Chloro(dimethyl)(phenyl)silane
CAS: 768-33-2; EC No: 212-193-0; MDL No: MFCD00000499
UN No: UN2987; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: C8H11ClSi; MW: 170.72
Flash point: 62°C (144°F)
Density (g/mL): 1.017; Refractive Index: 1.509
Moisture Sensitive
- Refractive index: 1.509
- Moisture Sensitive
- Liquid
Formula:
C₈H₁₁ClSi MW: 170.71 g/mol Density: 1.017 g/mL Flash Pt: 62 °C (144 °F) |
MDL Number:
MFCD00000499 CAS Number: 768-33-2 EINECS: 212-193-0 UN: 2987 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...