Quinoline ≥97%

Supplier: BeanTown Chemical

218615-100G 218615-2.5KG 218615-500G
BT218615-100GEA 45.21 USD
BT218615-100G BT218615-2.5KG BT218615-500G
Quinoline ≥97%
Quinoline

CAS: 91-22-5; EC No: 202-051-6; MDL No: MFCD00006736; RTECS: VA9275000 UN No: UN2656; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C9H7N; MW: 129.16 Melting Point: -17- -13°; Boiling Point: 237°; Flash point: 101°C (214°F) Density (g/mL): 1.093; Refractive Index: 1.625 Hygroscopic , Light Sensitive


  • RTECS: VA9275000
  • Refractive index: 1.625
  • Hygroscopic, Light Sensitive
  • Liquid

Formula: C₉H₇N
MW: 129.16 g/mol
Boiling Pt: 237 °C
Melting Pt: –17– –13 °C
Density: 1.093 g/mL
Flash Pt: 101 °C (214 °F)
MDL Number: MFCD00006736
CAS Number: 91-22-5
EINECS: 202-051-6
UN: 2656
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e452fdc6c4f70d31bc2cfb5dade041787f8be427&vid=ec752804bedb2e9f8adfbea9ad287c2452f87a17&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR