Potassium iodate ≥98%

Supplier: BeanTown Chemical

218390-100G 218390-2.5KG 218390-500G
BT218390-100GEA 45 USD
BT218390-100G BT218390-2.5KG BT218390-500G
Potassium iodate ≥98%
Potassium iodate

CAS: 7758-05-6; EC No: 231-831-9; MDL No: MFCD00011406; RTECS: NN1350000 UN No: UN1479; Haz Class: 5.1; Packing Group: II Crystalline/Powder; Linear Formula: KIO3; MW: 214.00 Melting Point: 560° Density (g/mL): 3.93


  • RTECS: NN1350000
  • Crystalline/Powder

Formula: KlO₃
Melting Pt: 560 °C
Density: 3.93 g/mL
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR