2-Chloro-2-methylhexane ≥97%
Supplier: BeanTown Chemical
218035-25G
218035-5G
BT218035-25GEA
192.91
USD
BT218035-25G
BT218035-5G
2-Chloro-2-methylhexane ≥97%
2-Chloro-2-methylhexane
CAS: 4398-65-6; MDL No: MFCD00060779
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH3(CH2)3CCl(CH3)2; Molecular Formula: C7H15Cl; MW: 134.65
- Liquid
Formula:
C₇H₁₅Cl |
MDL Number:
MFCD00060779 CAS Number: 4398-65-6 ADR: 3,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...