Zirconium(IV) butoxide ≥80% (w/w) in 1-butanol
Supplier: BeanTown Chemical
Synonyms:
Zirconium(IV) n-butoxide, Zirconium(IV) Tetrabutoxide
217550-1KG
217550-250G
217550-50G
BT217550-1KGEA
172.01
USD
BT217550-1KG
BT217550-250G
BT217550-50G
Zirconium(IV) butoxide ≥80% (w/w) in 1-butanol
Zirconium(IV) butoxide
CAS: 1071-76-7; EC No: 213-995-3; MDL No: MFCD00048765
UN No: UN1120; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: Zr[O(CH2)3CH3]4; Molecular Formula: C16H36O4Zr; MW: 383.68
Flash point: 38°C (100°F)
Density (g/mL): 1.049; Refractive Index: 1.466
Moisture Sensitive
- Refractive index: 1.466
- Moisture Sensitive
- Liquid
Formula:
C₁₆H₃₆O₄Zr Density: 1.049 g/mL Flash Pt: 38 °C (100 °F) |
MDL Number:
MFCD00048765 CAS Number: 1071-76-7 EINECS: 213-995-3 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...