N,O-Bis(trimethylsilyl)trifluoroacetamide with 10% trimethylchlorosilane (contains 10% TMCS)

Supplier: BeanTown Chemical

217305-100G 217305-25G 217305-5G
BT217305-100GEA 468.21 USD
BT217305-100G BT217305-25G BT217305-5G
N,O-Bis(trimethylsilyl)trifluoroacetamide with 10% trimethylchlorosilane (contains 10% TMCS)
N,O-Bis(trimethylsilyl)trifluoroacetamide with 10% trimethylchlorosilane

CAS: 25561-30-2; MDL No: MFCD00008269 UN No: UN2924; Haz Class: 3(8); Packing Group: II Liquid; Linear Formula: CF3C[=NSi(CH3)3]OSi(CH3)3; Molecular Formula: C8H18F3NOSi2; MW: 257.40 Melting Point: -10°; Boiling Point: 145-147°; Flash point: -9°C (16°F) Density (g/mL): 0.97; Refractive Index: 1.384 Moisture Sensitive


  • Refractive index: 1.384
  • Moisture Sensitive
  • Liquid

Formula: C₈H₁₈F₃NOSi₂
Boiling Pt: 145…147 °C
Melting Pt: –10 °C
Density: 0.97 g/mL
Flash Pt: –9 °C (16 °F)
MDL Number: MFCD00008269
CAS Number: 25561-30-2
ADR: 3(8),II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR