Potassium perruthenate(VII) ≥97%
Supplier: BeanTown Chemical
Synonyms:
Potassium perruthenate
216370-1G
216370-5G
BT216370-1GEA
165
USD
BT216370-1G
BT216370-5G
Potassium perruthenate(VII) ≥97%
Potassium perruthenate(VII)
CAS: 10378-50-4; EC No: 233-835-6; MDL No: MFCD00061333
UN No: UN1479; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: KRuO4; MW: 204.17
Melting Point: 440° (decomposes)
- Crystalline
Formula:
C₃₂H₃₆N₂O₅ Melting Pt: 440 °C (dec.) |
MDL Number:
MFCD00061333 CAS Number: 10378-50-4 EINECS: 233-835-6 ADR: 5.1,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...