Sodium nitrate ≥99.0% ACS
Supplier: BeanTown Chemical
216355-1KG
216355-250G
BT216355-1KGEA
86.83
USD
BT216355-1KG
BT216355-250G
Sodium nitrate ≥99.0% ACS
Sodium nitrate
CAS: 7631-99-4; EC No: 231-554-3; MDL No: MFCD00011119; RTECS: WC5600000
UN No: UN1498; Haz Class: 5.1; Packing Group: III
Solid; Linear Formula: NaNO3; MW: 84.99
Melting Point: 306° (decomposes); Boiling Point: 380°
Density (g/mL): 2.261
- RTECS: WC5600000
- Solid
Formula:
NaNO₃ MW: 84.99 g/mol Boiling Pt: 380 °C Melting Pt: 306 °C (dec.) Density: 2.261 g/mL |
MDL Number:
MFCD00011119 CAS Number: 7631-99-4 EINECS: 231-554-3 UN: 1498 ADR: 5.1,III |
Specification Test Results
Assay | 99.0% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...