(±)-2-Methyl-3-pentanol ≥99%

Supplier: BeanTown Chemical

216165-10G 216165-50G
BT216165-10GEA 88.48 USD
BT216165-10G BT216165-50G
(±)-2-Methyl-3-pentanol ≥99%
(±)-2-Methyl-3-pentanol

CAS: 565-67-3; EC No: 209-286-3; MDL No: MFCD00004567; RTECS: SA7400000 UN No: UN2282; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3CH2CH(OH)CH(CH3)2; Molecular Formula: C6H14O; MW: 102.17 Boiling Point: 128°; Flash point: 46°C (115°F) Density (g/mL): 0.819; Refractive Index: 1.417


  • RTECS: SA7400000
  • Refractive index: 1.417
  • Liquid

Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 128
Melting Pt: -53
Density: 0.819 g/mL
Flash Pt: 46 °C (115 °F)
MDL Number: MFCD00004567
CAS Number: 565-67-3
EINECS: 209-286-3
UN: 2282
ADR: 3 III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR