Copper(II) bromide ≥99%
Supplier: BeanTown Chemical
Synonyms:
Cupric bromide, Cuprous bromide
215750-100G
215750-25G
215750-500G
BT215750-100GEA
54.78
USD
BT215750-100G
BT215750-25G
BT215750-500G
Copper(II) bromide ≥99%
Copper (II) bromide
CAS: 7789-45-9; EC No: 232-167-2; MDL No: MFCD00010970
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: CuBr2; MW: 223.36
Melting Point: 498°
Density (g/mL): 4.77
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
Br₂Cu MW: 223.35 g/mol Melting Pt: 498 °C Density: 4.77 g/mL |
MDL Number:
MFCD00010970 CAS Number: 7789-45-9 EINECS: 232-167-2 UN: 3260 ADR: 8,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...