Crystal violet ≥90% ACS

Supplier: BeanTown Chemical

Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet

215520-100G 215520-25G
BT215520-100GEA 80 USD
BT215520-100G BT215520-25G
Crystal violet ≥90% ACS
Crystal Violet

CAS: 548-62-9; EC No: 208-953-6; MDL No: MFCD00011750; RTECS: BO9000000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Molecular Formula: C25H30ClN3; MW: 407.99 Melting Point: 205° (decomposes)


  • RTECS: BO9000000
  • Powder

Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
Melting Pt: 205 °C (dec.)
MDL Number: MFCD00011750
CAS Number: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: 9 III
Merck Index: 13,04406

Order Now

Specification Test Results

Assay 90.0% min.
Loss on drying ≤7.5%
Visual transition pH 0.8-2.6, green-blue-violet
Sensitivity Passes test
Absorbance characteristics Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR