Gold(I) chloride ≥99.99%
Supplier: BeanTown Chemical
215005-1G
215005-250MG
215005-5G
BT215005-1GEA
232
USD
BT215005-1G
BT215005-250MG
BT215005-5G
Gold(I) chloride ≥99.99%
Gold (I) chloride
CAS: 10294-29-8; EC No: 233-655-8; MDL No: MFCD00046175
UN No: UN3260; Haz Class: 8; Packing Group: III
Powder; Molecular Formula: AuCl; MW: 232.42
Melting Point: 289° (decomposes)
Density (g/mL): 7.57
Moisture Sensitive
- Moisture Sensitive
- Powder
Formula:
AuCl Melting Pt: 289 °C (dec.) Density: 7.57 g/mL |
MDL Number:
MFCD00046175 CAS Number: 10294-29-8 EINECS: 233-655-8 UN: 3260 ADR: 8,III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...