Fullerene C70 ≥99%, powder
Supplier: BeanTown Chemical
Synonyms:
Buckminster-Fullerene C70, [5,6]-Fullerene-C70
214485-1G
214485-250MG
BT214485-1GEA
1253
USD
BT214485-1G
BT214485-250MG
Fullerene C70 ≥99%, powder
Fullerene C70
CAS: 115383-22-7; MDL No: MFCD00282904
UN No: UN1325; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: C; MW: 12.011
- Powder
Formula:
C₇₀ MW: 840.77 g/mol |
MDL Number:
MFCD00282904 CAS Number: 115383-22-7 UN: 1325 ADR: 4.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...