Sulfur ≥99.9995%, pieces
Supplier: BeanTown Chemical
214375-100G
214375-25G
214375-500G
BT214375-100GEA
216
USD
BT214375-100G
BT214375-25G
BT214375-500G
Sulfur ≥99.9995%, pieces
Sulphur
CAS: 7704-34-9; EC No: 231-722-6; MDL No: MFCD00085316; RTECS: WS4250000
UN No: UN1350; Haz Class: 4.1; Packing Group: III
Solid; Molecular Formula: S; MW: 32.07
Melting Point: 117-120°; Boiling Point: 444.7°
Density (g/mL): 2.07
- RTECS: WS4250000
- Solid
Formula:
S Boiling Pt: 444.7 °C Melting Pt: 117…120 °C Density: 2.07 g/mL |
MDL Number:
MFCD00085316 CAS Number: 7704-34-9 EINECS: 231-722-6 UN: 1350 ADR: 4.1,III |
Specification Test Results
Assay | 99.9995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...