Vanadium ≥99.5% (trace metals basis), powder -325 mesh

Supplier: BeanTown Chemical

214160-10G 214160-50G
BT214160-10GEA 127.02 USD
BT214160-10G BT214160-50G
Vanadium ≥99.5% (trace metals basis), powder -325 mesh
Vanadium

CAS: 7440-62-2; EC No: 231-171-1; MDL No: MFCD00011453; RTECS: YW1355000 UN No: UN3089; Haz Class: 4.1; Packing Group: II Powder; Molecular Formula: V; MW: 50.94 Melting Point: 1890°; Boiling Point: 3380° Density (g/mL): 6.11


  • RTECS: YW1355000
  • Powder

Formula: V
Boiling Pt: 3380 °C
Melting Pt: 1890 °C
Density: 6.11 g/mL
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR