Tetrachloroethylene ≥99%
Supplier: BeanTown Chemical
Synonyms:
Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene
213180-100ML
213180-2.5L
213180-500ML
BT213180-100MLEA
26.66
USD
BT213180-100ML
BT213180-2.5L
BT213180-500ML
Tetrachloroethylene ≥99%
Tetrachloroethylene
CAS: 127-18-4; EC No: 204-825-9; MDL No: MFCD00000834; RTECS: KX3850000
UN No: UN1897; Haz Class: 6.1; Packing Group: III
Liquid; Linear Formula: Cl2C=CCl2; MW: 165.83
Melting Point: -22°; Boiling Point: 120-121°
Density (g/mL): 1.623; Refractive Index: 1.505
- RTECS: KX3850000
- Refractive index: 1.505
- Liquid
Formula:
C₂Cl₄ MW: 165.83 g/mol Boiling Pt: 120…121 °C Melting Pt: –22 °C Density: 1.623 g/mL |
MDL Number:
MFCD00000834 CAS Number: 127-18-4 EINECS: 204-825-9 UN: 1897 ADR: 6.1,III |
|
Specification Test Results
Assay | 99% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...