6-Thioguanine ≥98%
Supplier: BeanTown Chemical
212935-1G
212935-5G
BT212935-1GEA
82.43
USD
BT212935-1G
BT212935-5G
6-Thioguanine ≥98%
6-Thioguanine
CAS: 154-42-7; EC No: 205-827-2; MDL No: MFCD00233553; RTECS: UP0740000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C5H5N5S; MW: 167.19
Melting Point:
- RTECS: UP0740000
- Powder
Formula:
C₅H₅N₅S Melting Pt: >300 °C |
MDL Number:
MFCD00233553 CAS Number: 154-42-7 EINECS: 205-827-2 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...