Neopentyl bromide ≥98%
Supplier: BeanTown Chemical
Synonyms:
2,2-Dimethyl-1-propyl bromide, 1-Bromo-2,2-dimethylpropane
212450-25G
212450-5G
BT212450-25GEA
200
USD
BT212450-25G
BT212450-5G
Neopentyl bromide ≥98%
Neopentyl bromide
CAS: 630-17-1; EC No: 211-132-5; MDL No: MFCD00000209
UN No: UN1993; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3)3CCH2Br; MW: 151.05
Boiling Point: 104-106°; Flash point: 6°C (42°F)
Density (g/mL): 1.199; Refractive Index: 1.436
- Refractive index: 1.436
- Liquid
Formula:
C₅H₁₁Br Boiling Pt: 104…106 °C Density: 1.199 g/mL Flash Pt: 6 °C (42 °F) |
MDL Number:
MFCD00000209 CAS Number: 630-17-1 EINECS: 211-132-5 UN: 1993 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...