Potassium nitrate ≥99%
Supplier: BeanTown Chemical
CAS: 7757-79-1; EC No: 231-818-8; MDL No: MFCD00011409; RTECS: TT3700000
UN No: UN1486; Haz Class: 5.1; Packing Group: III
Crystalline; Molecular Formula: KNO3; MW: 101.11
Melting Point: 334°; Boiling Point: 400° (decomposes)
Density (g/mL): 2.109
Hygroscopic
- RTECS: TT3700000
- Hygroscopic
- Crystalline
Formula:
KNO₃ Boiling Pt: 400 °C (dec.) Melting Pt: 334 °C Density: 2.109 g/mL |
MDL Number:
MFCD00011409 CAS Number: 7757-79-1 EINECS: 231-818-8 UN: 1486 ADR: 5.1,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...