Nickel(II) nitrate hexahydrate ≥98%
Supplier: BeanTown Chemical
212180-100G
212180-500G
BT212180-100GEA
28.11
USD
BT212180-100G
BT212180-500G
Nickel(II) nitrate hexahydrate ≥98%
Nickel (II) nitrate hexahydrate
CAS: 13478-00-7; EC No: 236-068-5; MDL No: MFCD00149805; RTECS: QR7300000
UN No: UN2725; Haz Class: 5.1; Packing Group: III
Crystalline; Linear Formula: Ni(NO3)2·6H2O; MW: 290.80
Melting Point: 56°; Boiling Point: 137°
Density (g/mL): 2.05
Hygroscopic
- RTECS: QR7300000
- Hygroscopic
- Crystalline
Formula:
Ni(NO₃)₂·6H₂O MW: 290.79 g/mol Boiling Pt: 137 °C Melting Pt: 56 °C Density: 2.05 g/mL |
MDL Number:
MFCD00149805 CAS Number: 13478-00-7 EINECS: 236-068-5 UN: 2725 ADR: 5.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...