N-Bromosuccinimide ≥99%

Supplier: BeanTown Chemical

211895-1KG 211895-250G 211895-5KG
BT211895-1KGEA 155.49 USD
BT211895-1KG BT211895-250G BT211895-5KG
N-Bromosuccinimide ≥99%
N-Bromosuccinimide

CAS: 128-08-5; EC No: 204-877-2; MDL No: MFCD00005510 UN No: UN3261; Haz Class: 8; Packing Group: III Powder; Molecular Formula: C4H4BrNO2; MW: 177.99 Melting Point: 175-180° (decomposes) Density (g/mL): 2.098 Light Sensitive, Moisture Sensitive


  • Light Sensitive, Moisture Sensitive
  • Powder

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR