Acetonitrile ≥99%
Supplier: BeanTown Chemical
CAS: 75-05-8; EC No: 200-835-2; MDL No: MFCD00001878; RTECS: AL7700000
UN No: UN1648; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3CN; MW: 41.05
Melting Point: -48°; Boiling Point: 81-82°; Flash point: 2°C (36°F)
Density (g/mL): 0.786; Refractive Index: 1.344
- RTECS: AL7700000
- Refractive index: 1.344
- Liquid
Formula:
H₃CCN Boiling Pt: 81…82 °C Melting Pt: –48 °C Density: 0.786 g/mL Flash Pt: 2 °C (36 °F) |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...