Ethyl formate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Formic acid ethyl ester
211805-100ML
211805-2.5L
211805-500ML
BT211805-100MLEA
31.83
USD
BT211805-100ML
BT211805-2.5L
BT211805-500ML
Ethyl formate ≥97%
Ethyl formate
CAS: 109-94-4; EC No: 203-721-0; MDL No: MFCD00003294; RTECS: LQ8400000
UN No: UN1190; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: HCOOCH2CH3; Molecular Formula: C3H6O2; MW: 74.08
Melting Point: -80°; Boiling Point: 52-54°; Flash point: -19°C (-2°F)
Density (g/mL): 0.921; Refractive Index: 1.359
- RTECS: LQ8400000
- Refractive index: 1.359
- Liquid
Formula:
C₃H₆O₂ MW: 74.08 g/mol Boiling Pt: 52…54 °C Melting Pt: –80 °C Density: 0.921 g/mL Flash Pt: –19 °C (–2 °F) |
MDL Number:
MFCD00003294 CAS Number: 109-94-4 EINECS: 203-721-0 UN: 1190 ADR: 3,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...