Sodium dichromate dihydrate

Supplier: BeanTown Chemical

211265-100G 211265-2KG 211265-500G
BT211265-100GEA 26 USD
BT211265-100G BT211265-2KG BT211265-500G
Sodium dichromate dihydrate
Sodium dichromate dihydrate

CAS: 7789-12-0; EC No: 234-190-3; MDL No: MFCD00149166; RTECS: HX7750000 UN No: UN3087; Haz Class: 5.1 (6.1); Packing Group: II Crystalline; Linear Formula: Na2Cr2O7·2H2O; MW: 298.00 Melting Point: 91° Density (g/mL): 2.350 Hygroscopic


  • RTECS: HX7750000
  • Hygroscopic
  • Crystalline

Formula: Na₂Cr₂O₇·2H₂O
Melting Pt: 91 °C
Density: 2.350 g/mL
MDL Number: MFCD00149166
CAS Number: 7789-12-0
EINECS: 234-190-3
UN: 3087
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR