Phosphinic acid (Hypophosphorous acid) 50% (w/w) in water
Supplier: BeanTown Chemical
211115-4X500G
211115-500G
BT211115-4X500EA
209
USD
BT211115-4X500
BT211115-500G
Phosphinic acid (Hypophosphorous acid) 50% (w/w) in water
Phosphinic acid (Hypophosphorous acid)
CAS: 6303-21-5; EC No: 228-601-5; MDL No: MFCD02183592
UN No: UN3264; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: H3PO2; MW: 66.00
Density (g/mL): 1.206
- Liquid
Formula:
H₃O₂P MW: 66 g/mol Density: 1.206 g/mL |
MDL Number:
MFCD02183592 CAS Number: 6303-21-5 EINECS: 228-601-5 UN: 3264 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...