4-Propylphenol ≥98%
Supplier: BeanTown Chemical
Synonyms:
p-Propylphenol, 4-n-Propylphenol
211080-100G
211080-25G
211080-5G
BT211080-100GEA
732
USD
BT211080-100G
BT211080-25G
BT211080-5G
4-Propylphenol ≥98%
4-Propylphenol
CAS: 645-56-7; EC No: 211-446-2; MDL No: MFCD00002395; RTECS: SM8610000
UN No: UN3145; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C9H12O ; MW: 136.19
Boiling Point: 232°; Flash point: 106°C (223°F)
Density (g/mL): 0.983; Refractive Index: 1.523
- RTECS: SM8610000
- Refractive index: 1.523
- Liquid
Formula:
C₉H₁₂O Boiling Pt: 232 °C Density: 0.983 g/mL Flash Pt: 106 °C (223 °F) |
MDL Number:
MFCD00002395 CAS Number: 645-56-7 EINECS: 211-446-2 UN: 3145 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...