2-Methyl-2-oxazoline ≥99%
Supplier: BeanTown Chemical
Synonyms:
2-Methyl-4,5-dihydrooxazole
145945-100G
145945-25G
BT145945-100GEA
75.17
USD
BT145945-100G
BT145945-25G
2-Methyl-2-oxazoline ≥99%
2-Methyl-2-oxazoline
CAS: 1120-64-5; EC No: 214-316-3; MDL No: MFCD00005298
UN No: UN1993; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C4H7NO ; MW: 85.10
Boiling Point: 109-111°; Flash point: 20°C (68°F)
Density (g/mL): 1.005; Refractive Index: 1.434
- Refractive index: 1.434
- Liquid
Formula:
C₄H₇NO MW: 85.11 g/mol Boiling Pt: 109…111 °C Density: 1.005 g/mL Flash Pt: 20 °C (68 °F) |
MDL Number:
MFCD00005298 CAS Number: 1120-64-5 EINECS: 214-316-3 ADR: 3 II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...