Zinc acetate dihydrate 98.0-101.0% ACS

Supplier: BeanTown Chemical

Synonyms: Acetic acid zinc salt dihydrate, Zinc (II) acetate dihydrate, Zinc diacetate dihydrate

145565-1KG 145565-250G 145565-5KG
BT145565-1KGEA 79.95 USD
BT145565-1KG BT145565-250G BT145565-5KG
Zinc acetate dihydrate 98.0-101.0% ACS
Zinc acetate dihydrate

CAS: 5970-45-6; EC No: 209-170-2; MDL No: MFCD00066961; RTECS: ZG8750000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: Zn(CH3COO)2·2H2O; MW: 219.50 Melting Point: 237° Density (g/mL): 1.84


  • RTECS: ZG8750000
  • Powder

Formula: (H₃CCOO)₂Zn·2H₂O
Melting Pt: 237 °C
Density: 1.84 g/mL
MDL Number: MFCD00066961
CAS Number: 5970-45-6
EINECS: 209-170-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 98.0-101.0%
pH of 5% solution at 25°C 6.0-7.0
Insoluble matter ≤0.005%
Chloride (Cl) ≤5ppm
Sulfate (SO4) ≤0.005%
Calcium (Ca) ≤0.005%
Magnesium (Mg) ≤0.005%
Potassium (K) ≤0.01%
Sodium (Na) ≤0.05%
Iron (Fe) ≤5ppm
Lead (Pb) ≤0.002%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR