Lead(II) carbonate ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Lead carbonat
145335-25G
145335-5G
BT145335-25GEA
143.98
USD
BT145335-25G
BT145335-5G
Lead(II) carbonate ≥99.999% (trace metals basis)
Lead (II) carbonate
CAS: 598-63-0; EC No: 209-943-4; MDL No: MFCD00064631
UN No: UN2291; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: PbCO3; MW: 267.20
- Powder
Formula:
PbCO₃ MW: 267.21 g/mol |
MDL Number:
MFCD00064631 CAS Number: 598-63-0 EINECS: 209-943-4 UN: 2291 ADR: 6.1,III |
Specification Test Results
Assay | 99.999% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...