Sanger's reagent ≥99%
Supplier: BeanTown Chemical
Synonyms:
1,3-Dinitro-4-fluorobenzene, 2,4-Dinitrofluorobenzene, 1-Fluoro-2,4-dinitrobenzene
145310-100G
145310-25G
145310-5G
BT145310-100GEA
226.59
USD
BT145310-100G
BT145310-25G
BT145310-5G
Sanger's reagent ≥99%
Sanger's reagent
CAS: 70-34-8; EC No: 200-734-3; MDL No: MFCD00007056; RTECS: CZ7800000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C6H3FN2O4; MW: 186.10
Melting Point: 25-27°; Boiling Point: 178°/25 mmHg; Flash point: 164°C (327°F)
Density (g/mL): 1.482; Refractive Index: 1.569
- RTECS: CZ7800000
- Refractive index: 1.569
- Solid
Formula:
C₆H₃FN₂O₄ MW: 186.1 g/mol Boiling Pt: 178 °C/25 mmHg Melting Pt: 25…27 °C Density: 1.482 g/mL Flash Pt: 164 °C (327 °F) |
MDL Number:
MFCD00007056 CAS Number: 70-34-8 EINECS: 200-734-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...