3,6-Dioxaoctamethylenediamine ≥97%

Supplier: BeanTown Chemical

Synonyms: Bis-NH2-PEG2, 2,2'-(Ethylenedioxy)bis(ethylamine), 1,8-Diamino-3,6-dioxaoctane, 1,2-Bis(2-aminoethoxy)ethane

145025-10G 145025-250G 145025-50G
BT145025-10GEA 27 USD
BT145025-10G BT145025-250G BT145025-50G
3,6-Dioxaoctamethylenediamine ≥97%
3,6-Dioxaoctamethylenediamine

CAS: 929-59-9; EC No: 213-203-6; MDL No: MFCD00040474 UN No: UN2735; Haz Class: 8; Packing Group: III Liquid; Linear Formula: NH2CH2CH2OCH2CH2OCH2CH2NH2; Molecular Formula: C6H16N2O2; MW: 148.20 Boiling Point: 105-109°/6 mmHg; Flash point: 121°C (250°F) Density (g/mL): 1.015; Refractive Index: 1.461 Hygroscopic


  • Refractive index: 1.461
  • Hygroscopic
  • Liquid

Formula: C₆H₁₆N₂O₂
MW: 148.20 g/mol
Boiling Pt: 105…109 °C/6 mmHg
Density: 1.015 g/mL
Flash Pt: 121 °C (250 °F)
MDL Number: MFCD00040474
CAS Number: 929-59-9
EINECS: 213-203-6
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR