Lutetium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.127 mm

Supplier: BeanTown Chemical

144230-25X25MM 144230-50X50MM
BT144230-25X25EA 447 USD
BT144230-25X25 BT144230-50X50
Lutetium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.127 mm
Lutetium

CAS: 7439-94-3; EC No: 231-103-0; MDL No: MFCD00011098 UN No: UN3178; Haz Class: 4.1; Packing Group: III Solid; Molecular Formula: Lu; MW: 174.97 Melting Point: 1663°; Boiling Point: 3402° Density (g/mL): 9.84


  • Solid

Formula: Lu
Boiling Pt: 3402 °C
Melting Pt: 1663 °C
Density: 9.84 g/mL
MDL Number: MFCD00011098
CAS Number: 7439-94-3
EINECS: 231-103-0
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR