Strontium hydroxide octahydrate, Technical Grade

Supplier: BeanTown Chemical

144180-2KG 144180-500G
BT144180-2KGEA 120.04 USD
BT144180-2KG BT144180-500G
Strontium hydroxide octahydrate, Technical Grade
Strontium hydroxide octahydrate

CAS: 1311-10-0; EC No: 242-367-1; MDL No: MFCD00150395 UN No: UN3262; Haz Class: 8; Packing Group: III Crystalline/Lump; Linear Formula: Sr(OH)2·8H2O; MW: 265.76 Density (g/mL): 1.9 Air Sensitive , Hygroscopic


  • Air Sensitive, Hygroscopic
  • Crystalline/Lump

Formula: H₂O₂Sr·8H₂O
Density: 1.9 g/mL
MDL Number: MFCD00150395
CAS Number: 1311-10-0
EINECS: 242-367-1
UN: 3262
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR