Barium nitrate ≥99.0% ACS
Supplier: BeanTown Chemical
143080-2KG
143080-500G
BT143080-2KGEA
159
USD
BT143080-2KG
BT143080-500G
Barium nitrate ≥99.0% ACS
Barium nitrate
CAS: 10022-31-8; EC No: 233-020-5; MDL No: MFCD00003442; RTECS: CQ9625000
UN No: UN1446; Haz Class: 5.1 (6.1); Packing Group: II
Crystalline; Linear Formula: Ba(NO3)2 ; MW: 261.34
Melting Point: 592° (decomposes)
Hygroscopic
- RTECS: CQ9625000
- Hygroscopic
- Crystalline
Formula:
Ba(NO₃)₂ Melting Pt: 592 °C (dec.) |
MDL Number:
MFCD00003442 CAS Number: 10022-31-8 EINECS: 233-020-5 UN: 1446 ADR: 5.1,II |
Specification Test Results
Assay | 99.0% min. |
pH of 5% solution at 25°C | 5.0-8.0 |
Insoluble matter | ≤0.01% |
Chloride (Cl) | ≤5ppm |
Calcium (Ca) | ≤0.05% |
Potassium (K) | ≤0.005% |
Sodium (Na) | ≤0.005% |
Strontium (Sr) | ≤0.05% |
Heavy metals (as Pb) | ≤5ppm |
Iron (Fe) | ≤2ppm |
Appearance | White crystalline powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...